Learn More
Cayman Chemical BIotInyl TyramIde 10mg
SDP
Catalog No. 502939353
Encompass_Preferred
A reagent used for tyramide signal amplification; has been used in immunohistochemistry, ELISA, Western blot, and in situ hybridization applications
Catalog No. 50-293-9353 Supplier Cayman Chemical Supplier No. 2799710 MG
A reagent used for tyramide signal amplification; has been used in immunohistochemistry, ELISA, Western blot, and in situ hybridization applications