Learn More
Medchemexpress LLC HY-125658 10mg Medchemexpress, Biotinyl tyramide CAS:41994-02-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12565810MG
Encompass_Preferred
Medchemexpress, HY-125658 10mg Biotinyl tyramide CAS:41994-02-9 Biotinyl tyramide is a biotin derivative used for tyramide signal amplification (TSA), as a reagent to amplify both immunohistochemical signals and in situ hybridization protocols[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9694