Learn More
Cayman Chemical BIotInyl TyramIde 50mg
SDP
Catalog No. 502940506
Encompass_Preferred
A reagent used for tyramide signal amplification; has been used in immunohistochemistry, ELISA, Western blot, and in situ hybridization applications
Catalog No. 50-294-0506 Supplier Cayman Chemical Supplier No. 2799750 MG
A reagent used for tyramide signal amplification; has been used in immunohistochemistry, ELISA, Western blot, and in situ hybridization applications