Learn More
Enzo Life Sciences Biotinyl-VAD(OMe)-FMK (1mg)
SDP
Catalog No. 502008316
Encompass_Preferred
Caspase inhibitor. Irreversible, cell permeable broad-spectrum caspase inhibitor. Alternative name: Caspase family inhibitor (fluoromethylketone). Formula: C30H49FN6O8S. MW: 672.8. Purity: ≥90% (TLC). Appearance: Off-white to brown powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. The stock solution is stable at -20°C for 6-8 months. Handling: Keep cool and dry.
Catalog No. 50-200-8316 Supplier Enzo Life Sciences Supplier No. ALX260098M001
Caspase inhibitor. Irreversible, cell permeable broad-spectrum caspase inhibitor. Alternative name: Caspase family inhibitor (fluoromethylketone). Formula: C30H49FN6O8S. MW: 672.8. Purity: ≥90% (TLC). Appearance: Off-white to brown powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. The stock solution is stable at -20°C for 6-8 months. Handling: Keep cool and dry.