Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01 & B2M HIV Gag (SLYNTVATL) Complex Protein, 200 μg (monomer, MALS verified)
Catalog No. 502370232
Encompass_Preferred
Biotinylated HLA-A*02:01/B2M complex loaded with the HIV Gag peptide SLYNTVATL, supplied as a lyophilized 200 μg preparation. Expressed in HEK293 cells, His- and Avi-tagged, and verified as monomeric by SEC-MALS for use in immunology assays and HLA-restricted T-cell studies.
  • Biotinylated for streptavidin-based capture and detection.
  • Monomeric form verified by SEC-MALS for reliable molecular characterization.
  • High purity: >95% by SDS-PAGE and SEC-MALS.
  • Lyophilized formulation with trehalose for long-term stability.
  • Includes Avi and His tags to enable immobilization and detection.
  • Contains SLYNTVATL peptide for HIV Gag-specific T-cell recognition studies.
Catalog No. 50-237-0232 Supplier ACROBiosystems Supplier No. HLHH82E8200UG
Biotinylated HLA-A*02:01/B2M complex loaded with the HIV Gag peptide SLYNTVATL, supplied as a lyophilized 200 μg preparation. Expressed in HEK293 cells, His- and Avi-tagged, and verified as monomeric by SEC-MALS for use in immunology assays and HLA-restricted T-cell studies.
  • Biotinylated for streptavidin-based capture and detection.
  • Monomeric form verified by SEC-MALS for reliable molecular characterization.
  • High purity: >95% by SDS-PAGE and SEC-MALS.
  • Lyophilized formulation with trehalose for long-term stability.
  • Includes Avi and His tags to enable immobilization and detection.
  • Contains SLYNTVATL peptide for HIV Gag-specific T-cell recognition studies.