Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems ACROBiosystems Biotinylated Human HLA-A*02:01 & B2M & MAGE-A4 Complex Protein (KVLEHVVRV), 25 μg (HLP-H82E5-25ug)

Catalog No. 502370242
Encompass_Preferred

Biotinylated human HLA-A*02:01 & B2M complexed with the MAGE-A4 peptide KVLEHVVRV. The protein is monomeric, MALS-verified, produced in HEK293 cells, and biotinylated via an Avitag for use in binding and immunological assays.

  • Available in 25 μg and 200 μg sizes.
  • Contains a polyhistidine tag and site-specific biotinylation via Avitag.
  • Produced in human 293 (HEK293) expression system.
  • Purity greater than 90% by SDS-PAGE and greater than 95% by SEC-MALS.
  • Calculated molecular weights: heavy chain ~36.3 kDa and B2M ~13.8 kDa; migrates ~42-45 kDa and ~15 kDa on SDS-PAGE.
  • Includes the MAGE-A4 epitope KVLEHVVRV for HLA-A2 presentation.
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw cycles.

Catalog No. 50-237-0242 Supplier ACROBiosystems Supplier No. HLPH82E525UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated human HLA-A*02:01 & B2M complexed with the MAGE-A4 peptide KVLEHVVRV. The protein is monomeric, MALS-verified, produced in HEK293 cells, and biotinylated via an Avitag for use in binding and immunological assays.

  • Available in 25 μg and 200 μg sizes.
  • Contains a polyhistidine tag and site-specific biotinylation via Avitag.
  • Produced in human 293 (HEK293) expression system.
  • Purity greater than 90% by SDS-PAGE and greater than 95% by SEC-MALS.
  • Calculated molecular weights: heavy chain ~36.3 kDa and B2M ~13.8 kDa; migrates ~42-45 kDa and ~15 kDa on SDS-PAGE.
  • Includes the MAGE-A4 epitope KVLEHVVRV for HLA-A2 presentation.
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.