Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human latent GDF-8 Protein, His, Avitag - recombinant human myostatin (Asn 24-Ser 375), 200 μg

Catalog No. NC3396047
Encompass_Preferred

Biotinylated Human latent GDF-8 Protein, His, Avitag is recombinant human latent myostatin (GDF-8) spanning Asn 24-Ser 375, expressed in HEK293 cells. The construct contains an N-terminal biotin acceptor peptide and a polyhistidine tag and is enzymatically biotinylated for site-specific labeling. The product is supplied lyophilized and has >90% purity.

  • Sequence: Asn 24-Ser 375 (Accession O14793-1).
  • Tags: biotin acceptor peptide and polyhistidine tag.
  • Expression system: human 293 (HEK293) cells.
  • Molecular weight: calculated 43.7 kDa; migrates as 40 kDa and 53-55 kDa on SDS-PAGE.
  • Purity: greater than 90% as determined by SDS-PAGE.
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Storage: lyophilized at -20°C to -70°C (stable 12 months); reconstituted material stable at -70°C for 3 months.
  • Available sizes: 25 μg, 200 μg.

Catalog No. NC3396047 Supplier ACROBiosystems Supplier No. GD8H82Q3200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated Human latent GDF-8 Protein, His, Avitag is recombinant human latent myostatin (GDF-8) spanning Asn 24-Ser 375, expressed in HEK293 cells. The construct contains an N-terminal biotin acceptor peptide and a polyhistidine tag and is enzymatically biotinylated for site-specific labeling. The product is supplied lyophilized and has >90% purity.

  • Sequence: Asn 24-Ser 375 (Accession O14793-1).
  • Tags: biotin acceptor peptide and polyhistidine tag.
  • Expression system: human 293 (HEK293) cells.
  • Molecular weight: calculated 43.7 kDa; migrates as 40 kDa and 53-55 kDa on SDS-PAGE.
  • Purity: greater than 90% as determined by SDS-PAGE.
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Storage: lyophilized at -20°C to -70°C (stable 12 months); reconstituted material stable at -70°C for 3 months.
  • Available sizes: 25 μg, 200 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.