Learn More
Anatrace BIOTINYLATED A8-35 100MG
Catalog No. NC1597059
Encompass
Biotinylated A8-35 100mg; CAS# 1423685-21-5; Solubility: up to 20% in water; Appearance: White Solid; ANATRACE BAM01-100MG.
Catalog No. NC1597059 Supplier Anatrace Supplier No. BAM01 100 MG
Biotinylated A8-35 100mg; CAS# 1423685-21-5; Solubility: up to 20% in water; Appearance: White Solid; ANATRACE BAM01-100MG.