Learn More
Vector Laboratories Goat Anti-Human IgG Antibody Biotinylated 1.5 mg
SDP
Supplier:  Vector Laboratories BA30001.5
Encompass
Vector Laboratories affinity-purified antibodies are prepared using proprietary immunization schedules that produce high affinity antibodies. The antibodies are then purified by affinity chromatography, and cross-reactivities that are likely to interfere with specific labeling are removed by solid phase adsorption techniques. The biotinylated secondary antibodies are conjugated to ensure the maximum degree of labeling without compromising the specificity or affinity of the antibody. These antibodies are subjected to rigorous quality control assays and can be used for tissue and cell staining, ELISAs, and blots. Biotinylated Goat Anti-Human IgG (H+L) is supplied in solution and recognizes both heavy and light chains. With some exceptions, the recommended dilution for most applications is 1:200.
Catalog No. NC9312297