Learn More
Jackson Immuno Research Labs Biotin-SP (long spacer) AffiniPure Donkey Anti-Rabbit IgG (H+L), .5ml
SDP
Catalog No. NC9745676
Encompass_Preferred
Biotin-SP-AffiniPure Donkey Anti-Rabbit IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rat,Shp Sr Prot)
Catalog No. NC9745676 Supplier Jackson Immuno Research Labs Supplier No. 711065152
Biotin-SP-AffiniPure Donkey Anti-Rabbit IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rat,Shp Sr Prot)