Learn More
American Research Products Inc Biotinylated AntiMMAE ab
Catalog No. 5000306307
Encompass
Biotinylated Anti-MMAE antibody [8B4] is a payload antibody produced in rabbit Available in 10 g size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized proteins are shipped at ambient temperature
Catalog No. 50-003-06307 Supplier American Research Products Inc Supplier No. HDBS1191
Biotinylated Anti-MMAE antibody [8B4] is a payload antibody produced in rabbit Available in 10 g size with 7 - 14 business days lead time Ships via gel pack Store at -20 C to -80 C for 12 months in lyophilized form After reconstitution if not intended for use within a month aliquot and store at -80 C (Avoid repeated freezing and thawing) Lyophilized proteins are shipped at ambient temperature