Learn More
American Research Products Inc Biotinylated human Fc gamma RIIIa / CD16a (176V) protein 250UG
Catalog No. NC3255820
Encompass
The sequence of the extracellular domain of human CD16a (Gly 17-Gln 208) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.
Catalog No. NC3255820 Supplier American Research Products Inc Supplier No. HUGR3AVB
The sequence of the extracellular domain of human CD16a (Gly 17-Gln 208) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.