Learn More
American Research Products Inc Biotinylated human Fc gamma RIIIb / CD16b (NA1) protein 250UG
Catalog No. NC3255817
Encompass
The sequence of the extracellular domain of human CD16b (Gly 17-Ser 200) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.
Catalog No. NC3255817 Supplier American Research Products Inc Supplier No. HUGR3B1B
The sequence of the extracellular domain of human CD16b (Gly 17-Ser 200) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.