Learn More
American Research Products Inc Biotinylated human Fc gamma RIIa / CD32a (167R) protein 250UG
Catalog No. NC3256646
Encompass
The sequence of the extracellular domain of human CD32a (Ala 36-Ile 218) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.
Catalog No. NC3256646 Supplier American Research Products Inc Supplier No. HUGR2ARB
The sequence of the extracellular domain of human CD32a (Ala 36-Ile 218) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.