Learn More
Haematologic Technologies Inc BIOTINYLATED CMK 1MG
Catalog No. 50883491
Encompass_Preferred
Inhibitor, Biotinylated Phe-Pro-Arg (FPR) - Chloromethylketone, 1 milligram, Use: Tri-peptide chloromethylketones are used to irreversibly inhibit various serine proteases, Research use only, supplied in 10mM HCl, store at -80 Celsius.
Catalog No. 50-883-491 Supplier Haematologic Technologies Inc Supplier No. BFPRCK06
Add to Cart
Add to Cart
Inhibitor, Biotinylated Phe-Pro-Arg (FPR) - Chloromethylketone, 1 milligram, Use: Tri-peptide chloromethylketones are used to irreversibly inhibit various serine proteases, Research use only, supplied in 10mM HCl, store at -80 Celsius.