Learn More
Medchemexpress LLC Biotinylated covalent Btk inhibitor (CC-292) | 1202758-21-1 | 99.9% | 961.18 g/mol | C48H68N10O9S | 10 MG
SDP
Catalog No. 5000214759
Encompass_Preferred
CNX-500 is a biotinylated probe consisting of a covalent Bruton's tyrosine kinase (BTK) inhibitor chemically linked to biotin; it retains potent inhibitory activity (reported IC50 ≈ 0.5 nM) and is intended for target engagement and affinity-based experiments.
  • Biotin tag enables affinity capture and pull-down assays.
  • Covalent BTK inhibitor provides irreversible labeling of the target.
  • High potency toward BTK (IC50 ~0.5 nM) for sensitive detection.
  • High purity reduces off-target background.
  • Suitable for biochemical and affinity-based proteomics applications.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-14759 Supplier Medchemexpress LLC Supplier No. HY10033810MG
CNX-500 is a biotinylated probe consisting of a covalent Bruton's tyrosine kinase (BTK) inhibitor chemically linked to biotin; it retains potent inhibitory activity (reported IC50 ≈ 0.5 nM) and is intended for target engagement and affinity-based experiments.
  • Biotin tag enables affinity capture and pull-down assays.
  • Covalent BTK inhibitor provides irreversible labeling of the target.
  • High potency toward BTK (IC50 ~0.5 nM) for sensitive detection.
  • High purity reduces off-target background.
  • Suitable for biochemical and affinity-based proteomics applications.
  • Stable under recommended storage conditions for long-term use.