Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus IL-2 R beta&IL-2 R alpha&IL-2 R gamma Protein, Fc,Avitag™&Fc,Avitag™
Catalog No. 502106434
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus IL-2RB&IL-2RA&IL-2RG, Fc,Avitag™&Fc,Avitag™ (ILG-C82F6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Thr 240 (IL-2RB) & Glu 22 - Glu 238 (IL-2RA) & Leu 23 - Pro 255 (IL-2RG) (Accession # Q38J85-1 (IL-2RB) & A0A2K5UYC9-1 (IL-2RA) & Q38JL2-1 (IL-2RG)).;Biotinylated Human IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 53.0 kDa (IL-2RB) and 78.5 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™) and subunit IL-2RA&IL-2RG is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™).
Catalog No. 50-210-6434 Supplier ACROBiosystems Supplier No. ILGC82F6200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus IL-2RB&IL-2RA&IL-2RG, Fc,Avitag™&Fc,Avitag™ (ILG-C82F6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Thr 240 (IL-2RB) & Glu 22 - Glu 238 (IL-2RA) & Leu 23 - Pro 255 (IL-2RG) (Accession # Q38J85-1 (IL-2RB) & A0A2K5UYC9-1 (IL-2RA) & Q38JL2-1 (IL-2RG)).;Biotinylated Human IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 53.0 kDa (IL-2RB) and 78.5 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™) and subunit IL-2RA&IL-2RG is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™).