Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus LILRB2 / CD85d / ILT4 Protein, His,Avitag™
Catalog No. 502106891
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Cynomolgus LILRB2, His,Avitag™ (LI2-C82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 (Accession # BAE87348.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 56.4 kDa.
Catalog No. 50-210-6891 Supplier ACROBiosystems Supplier No. LI2C82E325UG
Recombinant Protein;25UG;Biotinylated Cynomolgus LILRB2, His,Avitag™ (LI2-C82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 (Accession # BAE87348.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 56.4 kDa.