Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus LIV-1 / SLC39A6 Protein, His,Avitag™
Catalog No. 502106349
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus LIV-1, His,Avitag (LV1-C82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Ile 309 (Accession # XP_005586923.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 36.2 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Cynomolgus LIV-1, His,Avitag (Cat. No. LV1-C82E5) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-210-6349 Supplier ACROBiosystems Supplier No. LV1C82E5200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus LIV-1, His,Avitag (LV1-C82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Ile 309 (Accession # XP_005586923.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 36.2 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Cynomolgus LIV-1, His,Avitag (Cat. No. LV1-C82E5) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.1-4 ng/mL (QC tested).