Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus TSLP Protein, His,Avitag™ (MALS verified)
Catalog No. 502106876
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus TSLP, His,Avitag™ (TSP-C82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # A0A2K5TXV0-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.8 kDa | 11.7 kDa & 7.1 kDa.
Catalog No. 50-210-6876 Supplier ACROBiosystems Supplier No. TSPC82E3200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus TSLP, His,Avitag™ (TSP-C82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # A0A2K5TXV0-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.8 kDa | 11.7 kDa & 7.1 kDa.