Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Biotinylated-D-Lysine Tfa Salt

Catalog No. 90075110MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-075-110MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-075-110MG Supplier Sigma Aldrich Supplier No. 90075110MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Biotinylated D-lysine (BDL) can be used to conjugate a biotin handle into bacterial peptidoglycan (PG) for various applications. For example, Suzanne Walker and coworkers reported that penicillin-binding protein PBP4 of Staphylococcus aureus could use BDL to biotinylate Lipid I and Lipid II, facilitating assays to measure infamously low-abundance pools of cellular lipid-linked PG precursors. BDL provides a tool for studying other aspects of PG assembly including antibacterial development efforts.Detection of Lipid-Linked Peptidoglycan Precursors by Exploiting an Unexpected Transpeptidase Reaction

Specifications

CAS 2128748-50-3
Melting Point >300°C
Molecular Formula C51H39ClNiP2
Linear Formula C51H39ClNiP2
Quantity 100 mg
Synonym cis-((S)-BINAP)Ni(o-tolyl)Cl
Molecular Weight (g/mol) 807.95 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.