Learn More
Vector Laboratories Galanthus Nivalis (Snowdrop) Lectin (GNL) Biotinylated 2 mg
SDP
Catalog No. NC0665353
Encompass
Galanthus nivalis lectin (GNL), unlike most mannose-specific lectins, is not a metalloprotein and does not require Ca++ or Mn++ for binding. Binding is preferentially directed toward structures containing (α-1,3) mannose residues. Also in contrast to most mannose-binding lectins, GNL will not bind α-linked glucose. GNL binds to many viral glycoproteins and to rat and mouse IgM but not IgG. The only human serum protein reported to bind to this lectin is α2-macroglobulin. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC0665353 Supplier Vector Laboratories Supplier No. B12452
Add to Cart
Add to Cart
Galanthus nivalis lectin (GNL), unlike most mannose-specific lectins, is not a metalloprotein and does not require Ca++ or Mn++ for binding. Binding is preferentially directed toward structures containing (α-1,3) mannose residues. Also in contrast to most mannose-binding lectins, GNL will not bind α-linked glucose. GNL binds to many viral glycoproteins and to rat and mouse IgM but not IgG. The only human serum protein reported to bind to this lectin is α2-macroglobulin. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.