Learn More
Vector Laboratories Griffonia (Bandeiraea) Simplicifolia Lectin II (GSL II, BSL II) Biotinylated 2 mg
SDP
Catalog No. NC1471029
Encompass
Griffonia simplicifolia lectin II (GSL II) is a dimeric glycoprotein composed of two subunits of nearly identical size. Each subunit has disulfide-linked chains and a binding site for α- or β-linkedN-acetylglucosamine residues. Unlike other N-acetylglucosamine-specific lectins, increasing the number of N-acetylglucosamine residues beyond two does not improve affinity. GSL II is unique in its ability to recognize exclusively α- or β-linked N-acetylglucosamine residues on the nonreducing terminal of oligosaccharides. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC1471029 Supplier Vector Laboratories Supplier No. B12152
Griffonia simplicifolia lectin II (GSL II) is a dimeric glycoprotein composed of two subunits of nearly identical size. Each subunit has disulfide-linked chains and a binding site for α- or β-linkedN-acetylglucosamine residues. Unlike other N-acetylglucosamine-specific lectins, increasing the number of N-acetylglucosamine residues beyond two does not improve affinity. GSL II is unique in its ability to recognize exclusively α- or β-linked N-acetylglucosamine residues on the nonreducing terminal of oligosaccharides. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.