Learn More
ACROBiosystems ACROBiosystems Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120 Protein, His,Avitag™
Catalog No. 502106650
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120, His,Avitag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession # G4XFJ5-1 (E46G, T396A, A497T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObiosystems.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 56.7 kDa. The protein migrates as 90-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 5 μg/mL (100 μL/well) can bind Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120, His,Avitag (Cat. No. GP0-V182E6) with a linear range of 0.156-1.25 μg/mL (QC tested).
Catalog No. 50-210-6650 Supplier ACROBiosystems Supplier No. GP0V182E6200UG
Recombinant Protein;200UG;Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120, His,Avitag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession # G4XFJ5-1 (E46G, T396A, A497T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObiosystems.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 56.7 kDa. The protein migrates as 90-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 5 μg/mL (100 μL/well) can bind Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120, His,Avitag (Cat. No. GP0-V182E6) with a linear range of 0.156-1.25 μg/mL (QC tested).