Learn More
Kactus Biotinylated Human CCR5 Protein-Nanodisc (CR5-HM4N191B) 100UG
Catalog No. NC3660783
Encompass
Recombinant Biotinylated Human CCR5 Protein-Nanodisc is expressed from HEK293 with His tag and Avi tag at the C-terminus.It contains Met1-Leu352.
Catalog No. NC3660783 Supplier Kactus Supplier No. CR5HM4N191B
Recombinant Biotinylated Human CCR5 Protein-Nanodisc is expressed from HEK293 with His tag and Avi tag at the C-terminus.It contains Met1-Leu352.