Learn More
ACROBiosystems BIOTINYLATED HUM,CD8 ALPH,25UG
Supplier:  ACROBiosystems CDAH82E325UG
Encompass_Preferred
Biotinylated Human CD8 alpha / CD8A Protein, His,Avitag™.CD8A, CD8, Leu2, MAL, p32.Formulation;Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Catalog No. NC1818323