Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Her2 / ErbB2, ultra sensitivity (primary amine labeling, long spacer);HEK293;200UG;HE2,ERBB2,CD340,HER-2,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1,ERBB-2,MLN-19,TKR-1
Catalog No. 501017956
Encompass_Preferred
Recombinant Protein;Biotinylated Human Her2 / ErbB2, ultra sensitivity (primary amine labeling, long spacer);200UG;MABSol Biotinylated Human Her2 / ErbB2, Superior Sensitivity (HE2-H822R) is expressed from human HEK293 cells. It contains AA Thr 23 - Thr 652 (Accession # AAA75493). It is the biotinylated form of Human Her2 / ErbB2 Protein (Cat. No. HE2-H5225).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.2 kDa. The protein migrates as 95-110 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Trastuzumab at 3 g/mL (100 L/well) can bind Biotinylated Human Her2, His Tag (Cat. No. HE2-H822R) with a linear range of 0.1-3.1 ng/mL (QC tested).
Catalog No. 50-101-7956 Supplier ACROBiosystems Supplier No. HE2H822R200UG
Recombinant Protein;Biotinylated Human Her2 / ErbB2, ultra sensitivity (primary amine labeling, long spacer);200UG;MABSol Biotinylated Human Her2 / ErbB2, Superior Sensitivity (HE2-H822R) is expressed from human HEK293 cells. It contains AA Thr 23 - Thr 652 (Accession # AAA75493). It is the biotinylated form of Human Her2 / ErbB2 Protein (Cat. No. HE2-H5225).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.2 kDa. The protein migrates as 95-110 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Trastuzumab at 3 g/mL (100 L/well) can bind Biotinylated Human Her2, His Tag (Cat. No. HE2-H822R) with a linear range of 0.1-3.1 ng/mL (QC tested).