Learn More
ACROBiosystems ACROBiosystems Biotinylated Human 2B4 / CD244 / SLAMF4 Protein, Avitag™,His Tag (recommended for biopanning)
Supplier:  ACROBiosystems 2B4H82E925UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human 2B4, Avitag,His Tag (recommended for biopanning) (2B4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession # Q9BZW8-2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.4 kDa. The protein migrates as 40-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human 2B4, Avitag,His Tag (recommended for biopanning) (Cat. No. 2B4-H82E9) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0.3-5 ng/mL (QC tested).;Immobilized Biotinylated Human 2B4, Avitag,His Tag (recommended for biopanning)
Catalog No. 50-210-6202