Learn More
ACROBiosystems ACROBiosystems Biotinylated Human 4-1BB / TNFRSF9 (87-186) Protein, His,Avitag™
Catalog No. 502106397
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human 4-1BB (87-186), His,Avitag™ (41B-H82E4) is expressed from human 293 cells (HEK293). It contains AA Asp 87 - Gln 186 (Accession # Q07011-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 14.1 kDa.
Catalog No. 50-210-6397 Supplier ACROBiosystems Supplier No. 41BH82E4200UG
Recombinant Protein;200UG;Biotinylated Human 4-1BB (87-186), His,Avitag™ (41B-H82E4) is expressed from human 293 cells (HEK293). It contains AA Asp 87 - Gln 186 (Accession # Q07011-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 14.1 kDa.