Learn More
Amsbio LLC Biotinylated Human ACE2 / ACEH Protein, His,Avitagâ„¢ (MALS verified)
Supplier:  Amsbio LLC AMS.AC2H82E625UG
Encompass
Biotinylated Human ACE2, His,Avitag (AC2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 740 (Accession # Q9BYF1-1).
Catalog No. 50-204-1219