Learn More
ACROBiosystems ACROBiosystems Biotinylated Human ACE2 / ACEH Protein, His,Avitag™ (MALS verified)
Catalog No. 502107093
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human ACE2, His,Avitag (AC2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 611 (Accession # Q9BYF1-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 72.3 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H3) at 1 μg/mL (100 μL/well) can bind Biotinylated Human ACE2, His,Avitag (Cat. No. AC2-H82E8) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-210-7093 Supplier ACROBiosystems Supplier No. AC2H82E8200UG
Recombinant Protein;200UG;Biotinylated Human ACE2, His,Avitag (AC2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 611 (Accession # Q9BYF1-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 72.3 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H3) at 1 μg/mL (100 μL/well) can bind Biotinylated Human ACE2, His,Avitag (Cat. No. AC2-H82E8) with a linear range of 0.039-0.625 μg/mL (QC tested).