Learn More
ACROBiosystems ACROBiosystems Biotinylated Human ACE2 / ACEH Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems AC2H82E71MG
Encompass_Preferred
Recombinant Protein;1MG;Biotinylated Human ACE2, His,Avitag (AC2-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 740 (Accession # Q9BYF1-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 87.2 kDa. The protein migrates as 95-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S1 protein, His Tag (Cat. No. S1N-C52H3) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ACE2, His,Avitag (Cat. No. AC2-H82E7) with a linear range of 2-40 ng/mL (QC tested).
Catalog No. 50-210-7352