Learn More
ACROBiosystems Biotinylated Human Alpha-Synuclein Protein, His,Avitag™
Catalog No. 502054753
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Alpha-Synuclein, His,Avitag (ALN-H82H8) is expressed from E.coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.0 kDa. The protein migrates as 23 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-205-4753 Supplier ACROBiosystems Supplier No. ALNH82H825UG
Recombinant Protein;25UG;Biotinylated Human Alpha-Synuclein, His,Avitag (ALN-H82H8) is expressed from E.coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.0 kDa. The protein migrates as 23 kDa under reducing (R) condition (SDS-PAGE).