Learn More
ACROBiosystems ACROBiosystems Biotinylated Human B7-1 / CD80 Protein, Avitag™,His Tag (recommended for biopanning)
Catalog No. 502106363
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human B7-1, Avitag,His Tag (recommended for biopanning) (B71-H82E9) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # P33681-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 26.5 kDa. The protein migrates as 44-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD80 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-1, Avitag,His Tag (recommended for biopanning) (Cat. No. B71-H82E9) with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-210-6363 Supplier ACROBiosystems Supplier No. B71H82E925UG
Recombinant Protein;25UG;Biotinylated Human B7-1, Avitag,His Tag (recommended for biopanning) (B71-H82E9) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # P33681-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 26.5 kDa. The protein migrates as 44-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD80 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-1, Avitag,His Tag (recommended for biopanning) (Cat. No. B71-H82E9) with a linear range of 0.1-3 ng/mL (QC tested).