Learn More
ACROBiosystems ACROBiosystems Biotinylated Human B7-H5 / Gi24 / VISTA Protein, Fc,Avitag™
Catalog No. 502106389
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 47.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6389 Supplier ACROBiosystems Supplier No. B75H82F3200UG
Recombinant Protein;200UG;Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 47.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.