Learn More
ACROBiosystems Biotinylated Human B7-H6 / NCR3LG1 Protein, Fc,Avitag™
Catalog No. 502055039
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human B7-H6, Fc,Avitag (B76-H82Wb) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 262 (Accession # Q68D85-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.8 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NKp30, Fc Tag (Cat. No. NC3-H5259) at 10 μg/mL (100 μL/well) can bind Biotinylated Human B7-H6, Fc,Avitag (Cat. No. B76-H82Wb) with a linear range of 0.6-10 ng/mL (QC tested).
Catalog No. 50-205-5039 Supplier ACROBiosystems Supplier No. B76H82WB200UG
Recombinant Protein;200UG;Biotinylated Human B7-H6, Fc,Avitag (B76-H82Wb) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 262 (Accession # Q68D85-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.8 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NKp30, Fc Tag (Cat. No. NC3-H5259) at 10 μg/mL (100 μL/well) can bind Biotinylated Human B7-H6, Fc,Avitag (Cat. No. B76-H82Wb) with a linear range of 0.6-10 ng/mL (QC tested).