Learn More
ACROBiosystems ACROBiosystems Biotinylated Human B7-H7 / HHLA2 Protein, His,Avitag™
Catalog No. 502106808
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human B7-H7, His,Avitag™ (B77-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 40.7 kDa.
Catalog No. 50-210-6808 Supplier ACROBiosystems Supplier No. B77H82E325UG
Recombinant Protein;25UG;Biotinylated Human B7-H7, His,Avitag™ (B77-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 40.7 kDa.