Learn More
ACROBiosystems ACROBiosystems Biotinylated Human BLAME / SLAMF8 Protein, His,Avitag™
Catalog No. 502106486
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human BLAME, His,Avitag (BLE-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Asp 233 (Accession # Q9P0V8-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.1 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BLAME, His Tag at 2 μg/mL (100 μL/well) can bind Biotinylated Human BLAME, His,Avitag (Cat. No. BLE-H82E4) with a linear range of 1-31 ng/mL (QC tested).
Catalog No. 50-210-6486 Supplier ACROBiosystems Supplier No. BLEH82E425UG
Recombinant Protein;25UG;Biotinylated Human BLAME, His,Avitag (BLE-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Asp 233 (Accession # Q9P0V8-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.1 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BLAME, His Tag at 2 μg/mL (100 μL/well) can bind Biotinylated Human BLAME, His,Avitag (Cat. No. BLE-H82E4) with a linear range of 1-31 ng/mL (QC tested).