Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Carbonic Anhydrase IX / CA9 (38-414) Protein, His,Avitag™ (MALS verified)
Catalog No. 502106537
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Carbonic Anhydrase IX (38-414), His,Avitag (CA9-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 44.5 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Carbonic Anhydrase IX (38-414), His,Avitag (Cat. No. CA9-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-Carbonic Anhydrase IX Antibody, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-210-6537 Supplier ACROBiosystems Supplier No. CA9H82E3200UG
Recombinant Protein;200UG;Biotinylated Human Carbonic Anhydrase IX (38-414), His,Avitag (CA9-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 44.5 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Carbonic Anhydrase IX (38-414), His,Avitag (Cat. No. CA9-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-Carbonic Anhydrase IX Antibody, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).