Learn More
ACROBiosystems BIOTINYLATED HUMAN CD19 PROTEI
Supplier:  ACROBiosystems CD9H825925UG
Encompass_Preferred
MABSol Biotinylated Human CD19, Fc Tag (CD9-H8259) is expressed from human HEK293 cells. It contains AA Pro 20 - Lys 291 (Accession # AAH06338). It is the biotinylated form of Human CD19 Protein, Fc Tag (Cat # CD9-H5259).
Catalog No. NC0850934