Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD200 / OX-2 Protein, Fc,Avitag™
Catalog No. 502106723
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD200, Fc,Avitag (OX2-H82F1) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # P41217-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.4 kDa. The protein migrates as 65-76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200 R1, His Tag (Cat. No. CR2-H52H6) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD200, Fc,Avitag (Cat. No. OX2-H82F1) with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-210-6723 Supplier ACROBiosystems Supplier No. OX2H82F125UG
Recombinant Protein;25UG;Biotinylated Human CD200, Fc,Avitag (OX2-H82F1) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # P41217-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.4 kDa. The protein migrates as 65-76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200 R1, His Tag (Cat. No. CR2-H52H6) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD200, Fc,Avitag (Cat. No. OX2-H82F1) with a linear range of 0.2-2 ng/mL (QC tested).