Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD23 / Fc epsilon RII Protein, His,Avitag™
Supplier:  ACROBiosystems CD3H82Q5200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD23, His,Avitag (CD3-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accession # P06734-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 34.2 kDa. The protein migrates as 40-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Immunoglobulin E, Human Plasma at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD23, His,Avitag (Cat. No. CD3-H82Q5) with a linear range of 0.156-2.5 μg/mL (QC tested).
Catalog No. 50-210-6551