Learn More
ACROBiosystems Biotinylated Human CD3E and CD3D Heterodimer Protein, Fc,His,Avitag and Fc,Flag,Avitag, 200 μg
Catalog No. 502055092
Encompass_Preferred
This biotinylated human CD3 epsilon and CD3 delta heterodimer protein is expressed in human HEK293 cells. The protein consists of CD3 epsilon and CD3 delta subunits, each containing a C-terminal human IgG1 Fc fragment, an Avitag™ and a tag (polyhistidine for CD3 epsilon; flag for CD3 delta). This complex is essential for T cell receptor-CD3 signaling pathways. The product is provided as a lyophilized powder, biotinylated enzymatically via the Avitag™ technology.
  • Greater than 90% purity by SDS-PAGE and greater than 95% purity by SEC-MALS
  • Verified molecular weight of 95-110 kDa by SEC-MALS
  • Validated for bioactivity via ELISA and BLI assays
  • Expressed in human HEK293 cells for authentic post-translational modifications
  • Supplied in a lyophilized format with trehalose protectant
Catalog No. 50-205-5092 Supplier ACROBiosystems Supplier No. CDDH82F6200UG
This biotinylated human CD3 epsilon and CD3 delta heterodimer protein is expressed in human HEK293 cells. The protein consists of CD3 epsilon and CD3 delta subunits, each containing a C-terminal human IgG1 Fc fragment, an Avitag™ and a tag (polyhistidine for CD3 epsilon; flag for CD3 delta). This complex is essential for T cell receptor-CD3 signaling pathways. The product is provided as a lyophilized powder, biotinylated enzymatically via the Avitag™ technology.
  • Greater than 90% purity by SDS-PAGE and greater than 95% purity by SEC-MALS
  • Verified molecular weight of 95-110 kDa by SEC-MALS
  • Validated for bioactivity via ELISA and BLI assays
  • Expressed in human HEK293 cells for authentic post-translational modifications
  • Supplied in a lyophilized format with trehalose protectant