Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD5 Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502106210
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD5, Fc,Avitag (CD5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 66.8 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6210 Supplier ACROBiosystems Supplier No. CD5H82F3200UG
Recombinant Protein;200UG;Biotinylated Human CD5, Fc,Avitag (CD5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 66.8 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.