Learn More
ACROBiosystems Biotinylated Human CD5 Protein, His,Avitag™
Catalog No. 502054886
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD5, His,Avitag (CD5-H82E5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 42.2 kDa.
Catalog No. 50-205-4886 Supplier ACROBiosystems Supplier No. CD5H82E5200UG
Recombinant Protein;200UG;Biotinylated Human CD5, His,Avitag (CD5-H82E5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 42.2 kDa.