Learn More
ACROBiosystems Biotinylated Human CD7 Protein, His,Avitag™
Supplier:  ACROBiosystems CD7H82E4200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD7, His,Avitag (CD7-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 20.0 kDa.
Catalog No. 50-205-4890