Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD7 Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502106196
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD7, Fc,Avitag (CD7-H82F3) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 44.6 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD7, Fc,Avitag (Cat. No. CD7-H82F3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-CD7 antibody, Mouse IgG1 with a linear range of 1-2 ng/mL (QC tested).
Catalog No. 50-210-6196 Supplier ACROBiosystems Supplier No. CD7H82F325UG
Recombinant Protein;25UG;Biotinylated Human CD7, Fc,Avitag (CD7-H82F3) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 44.6 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD7, Fc,Avitag (Cat. No. CD7-H82F3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-CD7 antibody, Mouse IgG1 with a linear range of 1-2 ng/mL (QC tested).