Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD72 Protein, His,Avitag™
Supplier:  ACROBiosystems CD2H82E3200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD72, His,Avitag (CD2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 31.7 kDa.
Catalog No. 50-210-6204