Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD79B Protein, His,Avitag™ (MALS verified)
Catalog No. 502107232
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD79B, His,Avitag (CDB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Accession # P40259-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.8 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD79B, His,Avitag (Cat. No. CDB-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-CD79B (Polatuzumab) Humanized Antibody with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-210-7232 Supplier ACROBiosystems Supplier No. CDBH82E325UG
Recombinant Protein;25UG;Biotinylated Human CD79B, His,Avitag (CDB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Accession # P40259-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.8 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD79B, His,Avitag (Cat. No. CDB-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-CD79B (Polatuzumab) Humanized Antibody with a linear range of 0.1-2 ng/mL (Routinely tested).