Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CEACAM-1 / CD66a Protein, Avitag™,His Tag
Catalog No. 502106594
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CEACAM-1, Avitag,His Tag (CE1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # P13688-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 45.9 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD209, His Tag (Cat. No. CD9-H5246) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-1, Avitag,His Tag (Cat. No. CE1-H82E5) with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-210-6594 Supplier ACROBiosystems Supplier No. CE1H82E5200UG
Recombinant Protein;200UG;Biotinylated Human CEACAM-1, Avitag,His Tag (CE1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # P13688-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 45.9 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD209, His Tag (Cat. No. CD9-H5246) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-1, Avitag,His Tag (Cat. No. CE1-H82E5) with a linear range of 10-78 ng/mL (QC tested).